siRNA Details
| Gene Symbol: Ica1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252266.1 | NP_001239195.1 | 10 | 1036 | ||
| NM_010492.3 | NP_034622.3 | 12 | 1512 | |||
| GenBank mRNA | AB303567.1 | - | 11 | 1023 | ||
| BC002030.1 | - | 12 | 1146 | 3487814 | ||
| CB056959.1 | - | 6 | 546 | |||
| DV658174.1 | - | 6 | 422 | |||
| U26459.1 | - | 12 | 1135 | |||
| U26460.1 | - | 11 | 1041 | |||
| U26461.1 | - | 10 | 977 | |||
| U34595.1 | - | 10 | 1008 | |||

Ica1
Genomic Map