siRNA Details
| Gene Symbol: Ica1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252266.1 | NP_001239195.1 | 6 | 749 | ||
| NM_010492.3 | NP_034622.3 | 7 | 1162 | |||
| GenBank mRNA | AB303567.1 | - | 6 | 673 | ||
| AK016989.1 | - | 7 | 865 | |||
| BC002030.1 | - | 7 | 796 | 3487814 | ||
| CB056959.1 | - | 2 | 259 | |||
| DV658174.1 | - | 1 | 72 | |||
| DV659030.1 | - | 5 | 534 | |||
| U26459.1 | - | 7 | 785 | |||
| U26460.1 | - | 6 | 691 | |||
| U26461.1 | - | 6 | 690 | |||
| U34595.1 | - | 5 | 658 | |||

Ica1
Genomic Map