siRNA Details
Gene Symbol: Id4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031166.2 | NP_112443.1 | 1 | 353 | ||
GenBank mRNA | AK041164.1 | - | 1 | 552 | ||
BC145367.1 | - | 2 | 275 | |||
X75018.1 | - | 1 | 353 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_175582.
Panther Classification:
Gene Ontology Categories:
