siRNA Details
| Gene Symbol: Ids | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010498.3 | NP_034628.2 | 6 | 1022 | ||
| GenBank mRNA | AJ627044.1 | - | 4 | 487 | ||
| AK085237.1 | - | 7 | 899 | |||
| AK132287.1 | - | 7 | 902 | |||
| AK166178.1 | - | 6 | 900 | |||
| AK166443.1 | - | 7 | 851 | |||
| L07921.1 | - | 6 | 899 | |||
| X75636.1 | - | 178 | ||||

Ids
Genomic Map