siRNA Details
| Gene Symbol: Idua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008325.4 | NP_032351.2 | 11 | 1842 | ||
| NM_172997.1 | NP_766585.1 | 10 | 1701 | |||
| GenBank mRNA | AK031414.1 | - | 10 | 1496 | ||
| AK045330.1 | - | 9 | 1329 | |||
| AK155627.1 | - | 11 | 1843 | |||
| AK169998.1 | - | 10 | 1500 | |||
| AK171124.1 | - | 6 | 853 | |||
| BC057935.1 | - | 10 | 1463 | 5327150 | ||
| L34111.1 | - | 10 | 1416 | |||

Idua
Genomic Map