siRNA Details
| Gene Symbol: Irgm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008326.1 | NP_032352.1 | 3 | 1151 | ||
| GenBank mRNA | AK002545.1 | - | 3 | 1173 | ||
| AK167558.1 | - | 2 | 1005 | |||
| AK171743.1 | - | 3 | 1132 | |||
| BC145957.1 | - | 3 | 963 | 40131260 | ||
| KC541561.1 | - | 1 | 658 | |||
| U19119.1 | - | 3 | 1151 | |||

Irgm1
Genomic Map