siRNA Details
| Gene Symbol: Ifi204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008329.2 | NP_032355.2 | 5 | 1079 | ||
| GenBank mRNA | BC010546.1 | - | 1057 | 4018506 | ||
| CT010351.1 | - | 971 | ||||
| M31419.1 | - | 1140 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifi204| Gene Symbol: Ifi204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008329.2 | NP_032355.2 | 5 | 1079 | ||
| GenBank mRNA | BC010546.1 | - | 1057 | 4018506 | ||
| CT010351.1 | - | 971 | ||||
| M31419.1 | - | 1140 | ||||