siRNA Details
| Gene Symbol: Ifi204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008329.2 | NP_032355.2 | 6, 7 | 1360 | ||
| GenBank mRNA | BC010546.1 | - | 1338 | 4018506 | ||
| BU694586.1 | - | 2, 3 | 282 | |||
| CT010351.1 | - | 1252 | ||||
| M31419.1 | - | 1421 | ||||

Ifi204
Genomic Map