siRNA Details
| Gene Symbol: Ifit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008332.3 | NP_032358.1 | 3 | 785 | ||
| GenBank mRNA | AK038027.1 | - | 3 | 538 | ||
| AK042366.1 | - | 3 | 567 | |||
| AK150206.1 | - | 2 | 387 | |||
| AK150253.1 | - | 2 | 387 | |||
| BC050835.1 | - | 3 | 533 | 6513417 | ||
| CT010395.1 | - | 1 | 309 | |||
| U43085.1 | - | 2 | 360 | |||

Ifit2
Genomic Map