siRNA Details
| Gene Symbol: Ifit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010501.2 | NP_034631.1 | 2 | 818 | ||
| GenBank mRNA | AK077459.1 | - | 2 | 817 | ||
| AK151032.1 | - | 2 | 818 | |||
| AK153226.1 | - | 2 | 819 | |||
| BC003804.1 | - | 2 | 784 | 3487345 | ||
| BC089563.1 | - | 2 | 816 | 30317428 | ||
| CT010266.1 | - | 1 | 747 | |||
| L32974.1 | - | 2 | 799 | |||
| Gene Symbol: I830012O16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005858.3 | NP_001005858.2 | 784 | |||
| GenBank mRNA | AK077243.1 | - | 773 | |||
| AK150632.1 | - | 2 | 1031 | |||
| AK151442.1 | - | 2 | 1031 | |||
| AK152620.1 | - | 2 | 1031 | |||

Ifit3
Genomic Map