siRNA Details
| Gene Symbol: HOXB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004502.3 | NP_004493.3 | 2 | 643 | ||
| GenBank mRNA | AK223249.1 | - | 2 | 642 | ||
| AK290653.1 | - | 2 | 609 | |||
| BC015345.1 | - | 2 | 590 | 4413080 | ||
| BM923808.1 | - | 2 | 644 | |||
| M16937.1 | - | 2 | 639 | |||
| M30598.1 | - | 2 | 186 | |||
| S49765.1 | - | 2 | 249 | |||

HOXB7
Genomic Map