siRNA Details
| Gene Symbol: Ifnar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010508.2 | NP_034638.2 | 11 | 1642 | ||
| GenBank mRNA | AK132431.1 | - | 11 | 1574 | ||
| AK136950.1 | - | 11 | 1618 | |||
| AK141630.1 | - | 11 | 1578 | |||
| AK148246.1 | - | 11 | 1584 | |||
| AK157555.1 | - | 12 | 1670 | |||
| AK178895.1 | - | 2 | 247 | |||
| BC043935.1 | - | 11 | 1593 | 4507009 | ||
| BC052217.1 | - | 11 | 1602 | 30008497 | ||
| BC052429.1 | - | 11 | 1604 | 6836300 | ||
| M89641.1 | - | 11 | 1607 | |||

Ifnar1
Genomic Map