siRNA Details
| Gene Symbol: Ifngr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010511.2 | NP_034641.1 | 6 | 956 | ||
| GenBank mRNA | AF128216.1 | - | 6 | 907 | ||
| AF128217.1 | - | 6 | 907 | |||
| AK087525.1 | - | 6 | 945 | |||
| AK150503.1 | - | 6 | 905 | |||
| AK151079.1 | - | 6 | 910 | |||
| AK152085.1 | - | 6 | 905 | |||
| AK152384.1 | - | 6 | 905 | |||
| AK153084.1 | - | 6 | 905 | |||
| AK153347.1 | - | 6 | 908 | |||
| AK159359.1 | - | 6 | 910 | |||
| AK159660.1 | - | 6 | 850 | |||
| AK159839.1 | - | 6 | 850 | |||
| AK169593.1 | - | 6 | 925 | |||
| AY157836.1 | - | 6 | 907 | |||
| AY157837.1 | - | 6 | 907 | |||
| BC132599.1 | - | 6 | 857 | 40130876 | ||
| BC138617.1 | - | 6 | 857 | 8861637 | ||
| J05265.1 | - | 6 | 933 | |||
| M25764.1 | - | 6 | 858 | |||
| M26711.1 | - | 6 | 933 | |||
| M28233.1 | - | 6 | 929 | |||
| M28995.1 | - | 6 | 834 | |||

Ifngr1
Genomic Map