siRNA Details
| Gene Symbol: Ifngr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008338.3 | NP_032364.1 | 6, 7 | 926 | ||
| GenBank mRNA | AK137165.1 | - | 6, 7 | 949 | ||
| AK143214.1 | - | 6, 7 | 941 | |||
| AK146995.1 | - | 944 | ||||
| AK165193.1 | - | 6, 7 | 942 | |||
| AK169907.1 | - | 6, 7 | 947 | |||
| BC055745.1 | - | 6, 7 | 926 | 6403454 | ||
| S69336.1 | - | 6, 7 | 950 | |||

Ifngr2
Genomic Map