siRNA Details
| Gene Symbol: Ifngr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008338.3 | NP_032364.1 | 5 | 752 | ||
| GenBank mRNA | AK137165.1 | - | 5 | 775 | ||
| AK143214.1 | - | 5 | 767 | |||
| AK146995.1 | - | 770 | ||||
| AK165193.1 | - | 5 | 768 | |||
| AK169907.1 | - | 5 | 773 | |||
| BC055745.1 | - | 5 | 752 | 6403454 | ||
| S69336.1 | - | 5 | 776 | |||

Ifngr2
Genomic Map