siRNA Details
| Gene Symbol: Ifrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013562.2 | NP_038590.2 | 7 | 993 | ||
| GenBank mRNA | AK078888.1 | - | 991 | |||
| AK168682.1 | - | 7 | 990 | |||
| AK215125.1 | - | 3 | 337 | |||
| AK219989.1 | - | 3 | 219 | |||
| BC037013.1 | - | 7 | 967 | |||
| BC043723.1 | - | 7 | 960 | 5369100 | ||
| X17400.1 | - | 7 | 970 | |||

Ifrd1
Genomic Map