siRNA Details
| Gene Symbol: Cd79b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008339.2 | NP_032365.1 | 5, 6 | 642 | ||
| GenBank mRNA | AK143573.1 | - | 5, 6 | 643 | ||
| BC012226.1 | - | 5, 6 | 642 | 4164551 | ||
| CT010358.1 | - | 5, 6 | 597 | |||
| J03857.1 | - | 5, 6 | 781 | |||

Cd79b
Genomic Map