siRNA Details
| Gene Symbol: Igf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122736.1 | NP_001116208.1 | 2 | 229 | ||
| NM_001122737.1 | NP_001116209.1 | 2 | 219 | |||
| NM_010514.3 | NP_034644.2 | 2 | 566 | |||
| GenBank mRNA | BB840527.1 | - | 2 | 220 | ||
| BC053489.1 | - | 2 | 205 | 30013295 | ||
| BC058615.1 | - | 2 | 204 | 5686066 | ||
| CF742200.1 | - | 2 | 425 | |||
| CN460564.1 | - | 2 | 441 | |||
| M14951.1 | - | 2 | 150 | |||

Igf2
Genomic Map