siRNA Details
| Gene Symbol: Igf2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010515.2 | NP_034645.2 | 40 | 6004 | ||
| GenBank mRNA | AK053045.1 | - | 5 | 596 | ||
| AK088699.1 | - | 15 | 2230 | |||
| BC031190.1 | - | 7 | 1010 | |||
| BC043457.1 | - | 3 | 205 | |||
| BC055018.1 | - | 3 | 210 | |||
| BC150817.1 | - | 40 | 5915 | 9087728 | ||
| BC171973.1 | - | 40 | 5915 | 9054259 | ||
| U04710.1 | - | 40 | 5960 | |||
| X60389.1 | - | 7 | 959 | |||

Igf2r
Genomic Map