siRNA Details
| Gene Symbol: Igfals | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008340.3 | NP_032366.2 | 2 | 751 | ||
| GenBank mRNA | AK004926.1 | - | 2 | 753 | ||
| AK220290.1 | - | 2 | 1263 | |||
| BC020357.1 | - | 2 | 771 | |||
| BC106111.1 | - | 2 | 754 | |||
| BC129875.1 | - | 2 | 739 | 40110389 | ||
| BC129876.1 | - | 2 | 739 | 40110391 | ||

Igfals
Genomic Map