siRNA Details
| Gene Symbol: Jchain | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152839.3 | NP_690052.2 | 4 | 543 | ||
| GenBank mRNA | AB644392.1 | - | 4 | 425 | ||
| AK078865.1 | - | 4 | 429 | |||
| AK160344.1 | - | 4 | 495 | |||
| BC006026.1 | - | 4 | 447 | 3491646 | ||
| M90766.1 | - | 4 | 446 | |||

Jchain
Genomic Map