siRNA Details
| Gene Symbol: Il10rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008349.5 | NP_032375.2 | 5, 6 | 730 | ||
| GenBank mRNA | AF440787.1 | - | 5, 6 | 657 | ||
| AK134042.1 | - | 5, 6 | 723 | |||
| AK160991.1 | - | 5, 6 | 1685 | |||
| AK181086.1 | - | 1, 2 | 107 | |||
| AK183290.1 | - | 1, 2 | 107 | |||
| AK196209.1 | - | 2, 3 | 217 | |||
| AK201410.1 | - | 1, 2 | 107 | |||
| BC145791.1 | - | 5, 6 | 730 | 40131094 | ||
| U53696.1 | - | 5, 6 | 643 | |||

Il10rb
Genomic Map