siRNA Details
| Gene Symbol: Il16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010551.3 | NP_034681.2 | 16, 17 | 3592 | ||
| GenBank mRNA | AF006001.1 | - | 3, 4 | 1322 | ||
| AF017111.1 | - | 4, 5 | 1472 | |||
| AF175292.1 | - | 16, 17 | 3592 | |||
| AK038319.1 | - | 4, 5 | 1508 | |||
| AK134007.1 | - | 4, 5 | 1423 | |||
| AK169551.1 | - | 4, 5 | 1495 | |||
| AK220455.1 | - | 16, 17 | 3602 | |||
| BC026894.1 | - | 4, 5 | 1401 | |||
| BC058709.1 | - | 16, 17 | 3625 | 6841305 | ||

Il16
Genomic Map