siRNA Details
| Gene Symbol: Il18rap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010553.4 | NP_034683.1 | 6, 7 | 1005 | ||
| GenBank mRNA | AF077347.1 | - | 6, 7 | 1014 | ||
| AK137541.1 | - | 6, 7 | 966 | |||
| BC120598.1 | - | 6, 7 | 1012 | 40129521 | ||
| BC120600.1 | - | 6, 7 | 1012 | 40129523 | ||

Il18rap
Genomic Map