siRNA Details
| Gene Symbol: Irak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177973.1 | NP_001171444.1 | 8 | 1158 | ||
| NM_001177974.1 | NP_001171445.1 | 8 | 1155 | |||
| NM_001177975.1 | NP_001171446.1 | 8 | 1155 | |||
| NM_001177976.1 | NP_001171447.1 | 8 | 1155 | |||
| NM_008363.2 | NP_032389.2 | 8 | 1155 | |||
| GenBank mRNA | AB088370.1 | - | 8 | 982 | ||
| AF103876.1 | - | 8 | 1155 | |||
| AK045958.1 | - | 8 | 1064 | |||
| AK052933.1 | - | 6 | 1025 | |||
| AK143694.1 | - | 8 | 1066 | |||
| AK144879.1 | - | 8 | 1064 | |||
| AY184362.1 | - | 8 | 1155 | |||
| AY184363.1 | - | 8 | 1155 | |||
| AY184364.1 | - | 8 | 1155 | |||
| BC004778.1 | - | 8 | 1150 | 3257579 | ||
| U56773.2 | - | 8 | 979 | |||

Irak1
Genomic Map