siRNA Details
| Gene Symbol: Il2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008367.3 | NP_032393.3 | 3 | 467 | ||
| GenBank mRNA | AK037369.1 | - | 3 | 453 | ||
| AK088594.1 | - | 3 | 409 | |||
| AK171665.1 | - | 3 | 404 | |||
| BC114437.1 | - | 3 | 373 | 40054387 | ||
| K02891.1 | - | 3 | 350 | |||
| M30856.1 | - | 3 | 397 | |||
| M54934.1 | - | 3 | 270 | |||

Il2ra
Genomic Map