siRNA Details
| Gene Symbol: HOXC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK000445.1 | - | 2 | 659 | ||
| AK315194.1 | - | 2 | 676 | |||
| BC032769.1 | - | 2 | 650 | |||
| BC053894.1 | - | 2 | 711 | 6144625 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008272, BC050838, AK078758, X07647, X55318, and others.

HOXC9
Genomic Map