siRNA Details
| Gene Symbol: HOXC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK000445.1 | - | 2 | 750 | ||
| AK315194.1 | - | 2 | 767 | |||
| BC032769.1 | - | 2 | 741 | |||
| BC053894.1 | - | 2 | 802 | 6144625 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXC9| Gene Symbol: HOXC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK000445.1 | - | 2 | 750 | ||
| AK315194.1 | - | 2 | 767 | |||
| BC032769.1 | - | 2 | 741 | |||
| BC053894.1 | - | 2 | 802 | 6144625 | ||