siRNA Details
| Gene Symbol: Il7r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008372.4 | NP_032398.3 | 4 | 547 | ||
| GenBank mRNA | AF078906.1 | - | 4 | 413 | ||
| AK040364.1 | - | 4 | 530 | |||
| AK040740.1 | - | 4 | 530 | |||
| AK041838.1 | - | 4 | 438 | |||
| AK042264.1 | - | 4 | 492 | |||
| AK154215.1 | - | 3 | 361 | |||
| AK154516.1 | - | 4 | 507 | |||
| BC089571.1 | - | 4 | 499 | 6758487 | ||
| M29697.1 | - | 4 | 460 | |||

Il7r
Genomic Map