siRNA Details
| Gene Symbol: Il7r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008372.4 | NP_032398.3 | 8 | 1092 | ||
| GenBank mRNA | AF078906.1 | - | 8 | 958 | ||
| AK040364.1 | - | 8 | 1075 | |||
| AK042264.1 | - | 8 | 1037 | |||
| AK154215.1 | - | 7 | 906 | |||
| AK154516.1 | - | 8 | 1053 | |||
| BC089571.1 | - | 8 | 1027 | 6758487 | ||

Il7r
Genomic Map