siRNA Details
| Gene Symbol: Fabp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008375.2 | NP_032401.1 | 2 | 229 | ||
| GenBank mRNA | BC119289.1 | - | 2 | 199 | 8734038 | |
| BC120767.1 | - | 2 | 199 | 40129690 | ||
| CJ043022.1 | - | 2 | 229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fabp6| Gene Symbol: Fabp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008375.2 | NP_032401.1 | 2 | 229 | ||
| GenBank mRNA | BC119289.1 | - | 2 | 199 | 8734038 | |
| BC120767.1 | - | 2 | 199 | 40129690 | ||
| CJ043022.1 | - | 2 | 229 | |||