siRNA Details
| Gene Symbol: Inpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008384.2 | NP_032410.2 | 5 | 813 | ||
| GenBank mRNA | AK078382.1 | - | 5 | 701 | ||
| BC025072.1 | - | 5 | 558 | 4456812 | ||
| U27295.1 | - | 5 | 662 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inpp1| Gene Symbol: Inpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008384.2 | NP_032410.2 | 5 | 813 | ||
| GenBank mRNA | AK078382.1 | - | 5 | 701 | ||
| BC025072.1 | - | 5 | 558 | 4456812 | ||
| U27295.1 | - | 5 | 662 | |||