siRNA Details
| Gene Symbol: Inpp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA881781.1 | - | 4 | 458 | ||
| AF040094.2 | - | 14 | 1908 | |||
| AK004601.1 | - | 15 | 1915 | |||
| AK004722.2 | - | 14 | 1810 | |||
| AY007563.1 | - | 21 | 2693 | |||
| BC028864.1 | - | 20 | 2626 | 4457437 | ||
| DV654857.1 | - | 3 | 478 | |||

Inpp5b
Genomic Map