siRNA Details
| Gene Symbol: Inpp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF040094.2 | - | 16, 17 | 2149 | ||
| AK004601.1 | - | 17, 18 | 2156 | |||
| AK004722.2 | - | 16, 17 | 2051 | |||
| AY007563.1 | - | 23, 24 | 2934 | |||
| BC028864.1 | - | 22, 23 | 2867 | 4457437 | ||
| BG802289.1 | - | 1, 2 | 142 | |||
| DV654857.1 | - | 5, 6 | 719 | |||

Inpp5b
Genomic Map