siRNA Details
| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010568.2 | NP_034698.2 | 7 | 2014 | ||
| GenBank mRNA | J05149.1 | - | 7 | 1573 | ||
| KC356871.1 | - | 7 | 1525 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Insr| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010568.2 | NP_034698.2 | 7 | 2014 | ||
| GenBank mRNA | J05149.1 | - | 7 | 1573 | ||
| KC356871.1 | - | 7 | 1525 | |||