siRNA Details
| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010568.2 | NP_034698.2 | 16 | 3631 | ||
| GenBank mRNA | J05149.1 | - | 16 | 3190 | ||
| KC356871.1 | - | 17 | 3178 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Insr| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010568.2 | NP_034698.2 | 16 | 3631 | ||
| GenBank mRNA | J05149.1 | - | 16 | 3190 | ||
| KC356871.1 | - | 17 | 3178 | |||