siRNA Details
| Gene Symbol: Invs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281978.1 | NP_001268907.1 | 6 | 858 | ||
| NM_010569.4 | NP_034699.3 | 5 | 742 | |||
| GenBank mRNA | AF034860.3 | - | 5 | 684 | ||
| AJ010902.1 | - | 5 | 683 | |||
| AK158766.1 | - | 6 | 817 | |||
| CK639743.1 | - | 2 | 221 | |||

Invs
Genomic Map