siRNA Details
| Gene Symbol: Irf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008391.4 | NP_032417.3 | 4 | 398 | ||
| GenBank mRNA | AK036652.1 | - | 4 | 366 | ||
| AK087926.1 | - | 4 | 384 | |||
| AK132099.1 | - | 4 | 411 | |||
| AK155025.1 | - | 4 | 383 | |||
| AK158642.1 | - | 2 | 114 | |||
| BC006577.1 | - | 4 | 391 | 3586586 | ||
| BC110666.1 | - | 4 | 370 | 4918447 | ||
| J03168.1 | - | 4 | 353 | |||

Irf2
Genomic Map