siRNA Details
| Gene Symbol: Irs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010571.3 | NP_034701.1 | 2 | 1585 | ||
| GenBank mRNA | AF035948.1 | - | 2 | 1391 | ||
| AK030311.1 | - | 2 | 1587 | |||
| AK040947.1 | - | 2 | 1570 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irs3| Gene Symbol: Irs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010571.3 | NP_034701.1 | 2 | 1585 | ||
| GenBank mRNA | AF035948.1 | - | 2 | 1391 | ||
| AK030311.1 | - | 2 | 1587 | |||
| AK040947.1 | - | 2 | 1570 | |||