siRNA Details
| Gene Symbol: Irf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159417.1 | NP_001152889.1 | 4 | 678 | ||
| NM_001159418.1 | NP_001152890.1 | 3 | 476 | |||
| NM_008394.3 | NP_032420.1 | 3 | 592 | |||
| GenBank mRNA | AK043697.1 | - | 1 | 53 | ||
| AK145911.1 | - | 2 | 344 | |||
| AK149742.1 | - | 2 | 343 | |||
| BC005435.1 | - | 4 | 453 | 3601255 | ||
| BC012968.1 | - | 3 | 487 | 3257714 | ||
| CK788642.1 | - | 3 | 419 | |||
| U51992.1 | - | 3 | 396 | |||

Irf9
Genomic Map