siRNA Details
| Gene Symbol: HOXC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017410.2 | NP_059106.2 | 1 | 533 | ||
| GenBank mRNA | AF255676.1 | - | 1 | 533 | ||
| AF263466.2 | - | 1 | 490 | |||
| AK024027.1 | - | 1 | 15 | |||
| BC002754.1 | - | 1 | 480 | |||
| BC090850.1 | - | 1 | 502 | 6171228 | ||
| BT009908.1 | - | 1 | 418 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010464, AF193796, AY349616.

HOXC13
Genomic Map