siRNA Details
| Gene Symbol: Itga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044268.1 | - | 2 | 157 | ||
| BC053031.1 | - | 20 | 2852 | 6401146 | ||
| BC062205.1 | - | 19 | 2627 | 4950774 | ||
| BC096741.1 | - | 21 | 2830 | |||
| D13867.1 | - | 20 | 2474 | |||

Itga3
Genomic Map