siRNA Details
| Gene Symbol: Itga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010576.3 | NP_034706.3 | 12 | 1526 | ||
| GenBank mRNA | AB850880.1 | - | 12 | 1291 | ||
| AF109136.1 | - | 12 | 1362 | |||
| AK051676.1 | - | 12 | 1452 | |||
| AK078842.1 | - | 12 | 1231 | |||
| AK089898.1 | - | 11 | 1326 | |||
| AK156015.1 | - | 12 | 1422 | |||
| AK156517.1 | - | 12 | 1296 | |||
| BC068313.1 | - | 12 | 1306 | 30093248 | ||
| X53176.1 | - | 12 | 1449 | |||

Itga4
Genomic Map