siRNA Details
| Gene Symbol: Itga5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010577.3 | NP_034707.3 | 17, 18 | 2009 | ||
| GenBank mRNA | AK142241.1 | - | 17, 18 | 1852 | ||
| AK144921.1 | - | 17, 18 | 1795 | |||
| AK148138.1 | - | 17, 18 | 1850 | |||
| AK200214.1 | - | 1, 2 | 107 | |||
| AK200582.1 | - | 1, 2 | 108 | |||
| BC050943.1 | - | 17, 18 | 2025 | 6821577 | ||
| BC053431.1 | - | 18, 19 | 2070 | |||
| BC059829.1 | - | 17, 18 | 2025 | 6821577 | ||
| X79003.1 | - | 17, 18 | 1966 | |||

Itga5
Genomic Map