siRNA Details
| Gene Symbol: Itga5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010577.3 | NP_034707.3 | 13 | 1534 | ||
| GenBank mRNA | AK142241.1 | - | 13 | 1377 | ||
| AK144921.1 | - | 13 | 1320 | |||
| AK148138.1 | - | 13 | 1375 | |||
| AK152594.1 | - | 13 | 1332 | |||
| AK211262.1 | - | 1 | 18 | |||
| AK218419.1 | - | 2 | 281 | |||
| BC050943.1 | - | 13 | 1550 | 6821577 | ||
| BC053431.1 | - | 14 | 1595 | |||
| BC059829.1 | - | 13 | 1550 | 6821577 | ||
| X79003.1 | - | 13 | 1491 | |||

Itga5
Genomic Map