siRNA Details
| Gene Symbol: Itgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010578.2 | NP_034708.1 | 12 | 1865 | ||
| GenBank mRNA | AK076526.1 | - | 8 | 1162 | ||
| AK088016.1 | - | 12 | 1767 | |||
| AK088729.1 | - | 12 | 1612 | |||
| AK135521.1 | - | 11 | 1541 | |||
| AK141253.1 | - | 7 | 792 | |||
| AK159689.1 | - | 12 | 1707 | |||
| AK167682.1 | - | 12 | 1763 | |||
| BC050906.1 | - | 12 | 1738 | 5721348 | ||
| X15202.1 | - | 11 | 1561 | |||
| Y00769.1 | - | 12 | 1653 | |||

Itgb1
Genomic Map