siRNA Details
| Gene Symbol: Itgb2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008405.3 | NP_032431.2 | 4 | 384 | ||
| GenBank mRNA | AF051367.1 | - | 4 | 369 | ||
| AK036640.1 | - | 4 | 384 | |||
| AK137534.1 | - | 4 | 385 | |||
| AK158108.1 | - | 4 | 385 | |||
| BC113142.1 | - | 4 | 353 | 40047633 | ||

Itgb2l
Genomic Map