siRNA Details
| Gene Symbol: Itpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010586.2 | NP_034716.1 | 38 | 5515 | ||
| NM_019923.4 | NP_064307.2 | 39 | 5614 | |||
| GenBank mRNA | AB182288.1 | - | 39 | 5303 | ||
| AB182290.1 | - | 38 | 5204 | |||
| BC025805.1 | - | 10 | 1404 | |||
| GU980658.1 | - | 39 | 5269 | |||
| Z71173.1 | - | 2 | 193 | |||

Itpr2
Genomic Map