siRNA Details
| Gene Symbol: Itpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010586.2 | NP_034716.1 | 8, 9 | 1205 | ||
| NM_019923.4 | NP_064307.2 | 9, 10 | 1304 | |||
| GenBank mRNA | AB182288.1 | - | 9, 10 | 993 | ||
| AB182290.1 | - | 8, 9 | 894 | |||
| AK030915.1 | - | 3, 4 | 329 | |||
| AK143206.1 | - | 8, 9 | 1233 | |||
| AK169446.1 | - | 8, 9 | 1161 | |||
| GU980658.1 | - | 9, 10 | 959 | |||

Itpr2
Genomic Map