siRNA Details
| Gene Symbol: Jag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010588.2 | NP_034718.2 | 26 | 3329 | ||
| GenBank mRNA | AF038572.1 | - | 26 | 3329 | ||
| BC009082.1 | - | 4 | 478 | |||
| Y14331.1 | - | 2 | 129 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jag2| Gene Symbol: Jag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010588.2 | NP_034718.2 | 26 | 3329 | ||
| GenBank mRNA | AF038572.1 | - | 26 | 3329 | ||
| BC009082.1 | - | 4 | 478 | |||
| Y14331.1 | - | 2 | 129 | |||